Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCA4 Monoclonal Antibody (5B7), Active Motif™

Rat Monoclonal Antibody
Supplier: Active Motif 61625
Description
Brg1 (Brahma-related gene 1) is an ATPase subunit of SWI2/SNF2-like chromatin-remodeling complexes that enable access of regulatory and effector proteins in transcription, DNA repair and DNA replication. Although Brg1-containing complexes are not essential for general cell survival, they participate in transcriptional regulation of several hundred genes including those involved in interferon and stress response, immune cells differentiation, neurogenesis, cell cycle etc. and is absolutely necessary for mouse embryogenesis. Brg1 is also involved in cell growth arrest, senescence and tumour supression.Specifications
| SMARCA4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| P51532, Q3TKT4 | |
| Smarca4 | |
| This antibody was raised against a recombinant protein corresponding to amino acids 213-295 of human SMARCA4. | |
| 100 μg | |
| Primary | |
| Human, Monkey, Mouse | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 5B7 | |
| Unconjugated | |
| Smarca4 | |
| ATP-dependent helicase SMARCA4; b2b508.1Clo; b2b692Clo; BAF190; BAF190A; brahma protein-like 1; Brg1; brg-1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; CSS4; FLJ39786; global transcription activator homologous sequence; homeotic gene regulator; HP1-BP72; hSNF2b; Mitotic growth and transcription activator; MRD16; nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1; RTPS2; SMARCA4; SNF2; SNF2B; SNF2beta; SNF2-beta; Snf2l4; SNF2LB; SNF2-like 4; sucrose nonfermenting-like 4; SW1/SNF; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SWI2; Transcription activator BRG1 | |
| Rat | |
| Protein G | |
| RUO | |
| 20586, 6597 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction