Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCA5 Monoclonal Antibody (14SNF-1G2)
GREENER_CHOICE

Catalog No. MA3055
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3055 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3055 Supplier Invitrogen™ Supplier No. MA3055
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA3-055 detects SNF2h in human samples. MA3-055 has been succesfully used in western blot application. Immunogen is a synthetic peptide corresponding to residues 1028-1045 of human SNF2h.

SMARCA5 is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. SMARCA5 is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein encoded by this gene is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The encoded protein is similar in sequence to the Drosophila ISWI chromatin remodeling protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCA5
Applications Western Blot
Classification Monoclonal
Clone 14SNF-1G2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.05% sodium azide
Gene SMARCA5
Gene Accession No. O60264
Gene Alias 4933427E24Rik; D030040M08Rik; D330027N15Rik; hISWI; hSNF2H; ISWI; MommeD4; mSnf2h; SMARCA5; SNF2H; sucrose nonfermenting protein 2 homolog; sucrose nonfermenting-like 5; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5; WCRF135
Gene Symbols SMARCA5
Host Species Mouse
Immunogen Synthetic peptide coupled to ovalbumin corresponding to amino acids 1028-1045 of human SNF2h.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8467
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.