Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCA6 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100278IR
Description
SMARCA6 Polyclonal antibody specifically detects SMARCA6 in Human samples. It is validated for Western BlotSpecifications
SMARCA6 | |
Polyclonal | |
Western Blot | |
EC 3.6.1, EC 3.6.4.-, helicase, lymphoid-specific, LSHSWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, Nbla10143, PASGFLJ10339, Proliferation-associated SNF2-like protein, SMARCA6lymphoid-specific helicase, subfamily A, member 6, SWI/SNF2-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 6 | |
The immunogen this antibody was made to, maps to a region between residues 550 and 600 of human Helicase, Lymphoid-Specific using the numbering given in entry NP_060533.2 (GeneID 3070). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
3070 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction