Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576839 Supplier Invitrogen™ Supplier No. PA576839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

SMARCAD1 belongs to the SNF2/RAD54 helicase family. It contains 2 CUE domains, 1 helicase ATP-binding domain, and 1 helicase C-terminal domain. It is a probable ATP-dependent DNA helicase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCAD1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SMARCAD1
Gene Accession No. D3Z9Z9, Q04692, Q9H4L7
Gene Alias ADERM; ATP-dependent helicase 1; ATP-dependent helicase SMARCAD1; AV081750; AW226546; D6Pas1; DKFZP762K2015; enhancer trap locus 1; enhancer trap locus homolog 1; Etl1; Etl-1; HEL1; hHEL1; KIAA1122; mKIAA1122; Smarcad1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A containing DEAD/H box 1; SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1
Gene Symbols SMARCAD1
Host Species Rabbit
Immunogen Recombinant full length Human SMARCAD1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13990, 312398, 56916
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.