Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SmarcAL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SmarcAL1 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SmarcAL1 Polyclonal specifically detects SmarcAL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SmarcAL1 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
50485 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: SYELGQGHAQASPEIRFTPFANPTHKPLAKPKSSQETPAHSSGQPPRDAKLEAKTAKASPSGQNISYIHSSSESVTPRTEGRLQQKSGSSVQKGVNSQKGKCVRNGDRFQVLIGYNAELIAVFKTLPSKNYDPDTKTWNFS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 - 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Rabbit | |
Human | |
ATP-driven annealing helicase, EC 3.6.1, EC 3.6.4.-, HARPSWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A-like protein 1, HepA-related protein, HHARP, SIOD, SMARCA-like protein 1, subfamily a-like 1, Sucrose nonfermenting protein 2-like 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin | |
SMARCAL1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title