Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SmarcAL1 Antibody (PCRP-SMARCAL1-1C4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32055520UG
Description
SmarcAL1 Monoclonal antibody specifically detects SmarcAL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SmarcAL1 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
ATP-driven annealing helicase, EC 3.6.1, EC 3.6.4.-, HARPSWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A-like protein 1, HepA-related protein, HHARP, SIOD, SMARCA-like protein 1, subfamily a-like 1, Sucrose nonfermenting protein 2-like 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin | |
Recombinant full-length human SmarcAL1 protein | |
20 μg | |
Chromatin Modifiers | |
50485 | |
Store at 4&drg;C. Do not freeze. | |
IgG2c κ |
Immunohistochemistry (Paraffin) | |
PCRP-SMARCAL1-1C4 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction