Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCAL1 / HARP Monoclonal Antibody (4B10), Active Motif™
SDP

Catalog No. 501993155
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3155 100 μg
50-199-3156 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3155 Supplier Active Motif Supplier No. 61115
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRKCQ is a calcium-independent, novel isoform that is important in transducing signals in T lymphocytes. Interaction of a T lymphocyte with an antigen-presenting cell results in clustering of the T cell antigen receptor (TCR) and the recruitment of a large signaling complex to the site of cell-cell contact. PRKCQ is essential for TCR-mediated activation. PKC theta plays a role in pathways that link the TCR signaling complex to the activation of NF-kappa B in mature T lymphocytes.

Specifications

Antigen SMARCAL1 / HARP
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene SMARCAL1
Gene Accession No. Q8BJL0, Q9NZC9, Q9TTA5
Gene Alias 6030401P21Rik; AI851716; ATP-driven annealing helicase; Harp; HepA-related protein; hHARP; mharp; Smarcal1; SMARCA-like protein 1; Sucrose nonfermenting protein 2-like 1; Swi/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1
Gene Symbols SMARCAL1
Host Species Mouse
Immunogen This SMARCAL1 / HARP antibody was raised against purifed bovine SMARCAL1 / HARP protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338072, 50485, 54380
Target Species Bovine, Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.