Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554181 Supplier Invitrogen™ Supplier No. PA554181
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYELGQGHAQ ASPEIRFTPF ANPTHKPLAK PKSSQETPAH SSGQPPRDAK LEAKTAKASP SGQNISYIHS SSESVTPRTE GRLQQKSGSS VQKGVNSQKG KCVRNGDRFQ VLIGYNAELI AVFKTLPSKN YDPDTKTWNF S Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 39%.

PRKCQ is a calcium-independent, novel isoform that is important in transducing signals in T lymphocytes. Interaction of a T lymphocyte with an antigen-presenting cell results in clustering of the T cell antigen receptor (TCR) and the recruitment of a large signaling complex to the site of cell-cell contact. PRKCQ is essential for TCR-mediated activation. PKC theta plays a role in pathways that link the TCR signaling complex to the activation of NF-kappa B in mature T lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCAL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMARCAL1
Gene Accession No. Q9NZC9
Gene Alias 6030401P21Rik; AI851716; ATP-driven annealing helicase; DNA-dependent ATPase A; HARP; HepA-related protein; HHARP; Mharp; SMARCAL1; SMARCA-like protein 1; Sucrose nonfermenting protein 2-like 1; Swi/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1
Gene Symbols SMARCAL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMARCAL1. Recombinant protein control fragment (Product #RP-91637).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50485
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.