Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113467
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113467 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113467 Supplier Invitrogen™ Supplier No. PA5113467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMARCAL1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

PRKCQ is a calcium-independent, novel isoform that is important in transducing signals in T lymphocytes. Interaction of a T lymphocyte with an antigen-presenting cell results in clustering of the T cell antigen receptor (TCR) and the recruitment of a large signaling complex to the site of cell-cell contact. PRKCQ is essential for TCR-mediated activation. PKC theta plays a role in pathways that link the TCR signaling complex to the activation of NF-kappa B in mature T lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCAL1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SMARCAL1
Gene Accession No. Q8BJL0, Q9NZC9
Gene Alias 6030401P21Rik; AI851716; ATP-driven annealing helicase; DNA-dependent ATPase A; HARP; HepA-related protein; HHARP; Mharp; SMARCAL1; SMARCA-like protein 1; Sucrose nonfermenting protein 2-like 1; Swi/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1
Gene Symbols SMARCAL1
Host Species Rabbit
Immunogen Recombinant Human SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1 protein (6-277AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50485, 54380
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.