Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCB1 Recombinant Rabbit Monoclonal Antibody (JB49-18)
GREENER_CHOICE

Catalog No. PIMA541260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541260 Supplier Invitrogen™ Supplier No. MA541260
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

SMARCB1 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

The SWI-SNF complex is involved in the activation of transcription via the remodeling of nucleosome structure in an ATP-dependent manner. Brm (also designated SNF2alpha) and Brg-1 (also designated SNF2beta) are the ATPase subunits of the mammalian SWI-SNF complex. Brm, Brg-1, Ini1 (integrase interactor 1, also designated SNF5), BAF155 (also designated SRG3) and BAF170 are thought to comprise the functional core of the SWI-SNF complex. Addition of Ini1, BAF155 and BAF170 to Brg-1 appears to increase remodeling activity. Other complex subunits are thought to play regulatory roles. hSNF2L and hSNF2H both appear to be homologs of Drosophila ISWI, a Brm related ATPase that is present in chromatin remodeling complexes other than SWI/SNF, including the NURF (nucleosome remodeling factor).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB49-18
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene SMARCB1
Gene Accession No. Q12824, Q9Z0H3
Gene Alias AU020204; Baf47; BRG1-associated factor 47; BRG1-associated factor 47 (BAF47); CSS3; hSNF5; hSNFS; INI1; Integrase interactor 1 protein; malignant rhabdoid tumor suppressor; MRD15; mSNF5; PPP1R144; protein phosphatase 1, regulatory subunit 144; RDT; RTPS1; Sfh1p; SMARCB1; smarcb1a; SNF5; SNF5 homolog; SNF5/INI1; SNF5L1; Snr1; Sucrose nonfermenting yeast homolog like 1; sucrose nonfermenting, yeast, homolog-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 isoform a; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1a; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1-A; SWI/SNF-related matrix-associated protein; SWI10; SWNTS1; Transcription factor TYE4; Transcription regulatory protein SNF5; TYE4; Unknown (protein for MGC:138029); zgc:92517
Gene Symbols SMARCB1
Host Species Rabbit
Immunogen Synthetic peptide of C terminal Human SNF5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20587, 361825, 6598
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.