Learn More
SMARCB1/SNF5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | SMARCB1/SNF5 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1718
![]() |
Bethyl Laboratories, Inc
IHC00228T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1717
![]() |
Bethyl Laboratories, Inc
IHC00228 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_101
The protein encoded by this gene is part of a complex that relieves repressive chromatin structures, allowing the transcriptional machinery to access its targets more effectively. The encoded nuclear protein may also bind to and enhance the DNA joining activity of HIV-1 integrase. This gene has been found to be a tumor suppressor, and mutations in it have been associated with maligt rhabdoid tumors. Two transcript variants encoding different isoforms have been found for this gene.Specifications
SMARCB1/SNF5 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
BAF47, INI1, RDT, SNF5, SNF5L1, Sfh1p, Snr1, hSNFS | |
Between 75 and 125 | |
Primary | |
4° C | |
SMARCB1 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q12824, Q9Z0H3 | |
20587, 6598 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.