Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCC1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298777AF405
Description
SMARCC1 Polyclonal specifically detects SMARCC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SMARCC1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
6599 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SMARCC1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction