Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCC1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298776JF646
Description
SMARCC1 Polyclonal antibody specifically detects SMARCC1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SMARCC1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SMARCC1. | |
0.1 mL | |
Chromatin Modifiers | |
6599 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction