Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCC1 Antibody (PCRP-SMARCC1-1F1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308611APC
Description
SMARCC1 Monoclonal antibody specifically detects SMARCC1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
SMARCC1 | |
Monoclonal | |
APC | |
BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3 | |
Recombinant full-length human SMARCC1 protein (Uniprot: Q92922) | |
100 μg | |
Chromatin Modifiers | |
6599 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, ELISA, Immunofluorescence, Protein Array | |
PCRP-SMARCC1-1F1 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction