Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCC1 Rabbit anti-Human, Mouse, Clone: 1R7I6, Novus Biologicals™
SDP

Catalog No. p-7247834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167826 20 μg
NB167825 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167826 Supplier Novus Biologicals Supplier No. NBP31636720UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SMARCC1 Monoclonal antibody specifically detects SMARCC1 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunofluorescence, Immunoprecipitation

Specifications

Antigen SMARCC1
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 1R7I6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 700-800 of human SMARCC1 (Q92922).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 6599
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.