Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCC1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $524.00
Specifications
Antigen | SMARCC1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SMARCC1 Polyclonal specifically detects SMARCC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
SMARCC1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
6599 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown | |
Unconjugated | |
Rabbit | |
Human | |
BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 80-320 of human SMARCC1 (NP_003065.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title