Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079836
Description
SMARCD1 Polyclonal specifically detects SMARCD1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
SMARCD1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A, BRG1-associated factor 60A, chromatin remodeling complex BAF60A subunit, CRACD1, Rsc6p, subfamily d, member 1, SWI/SNF complex 60 kDa subunit, SWI/SNF complex 60 kDa subunit A, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 1,60 kDa BRG-1/Brm-associated factor subunit A, Swp73-like protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
Q96GM5 | |
SMARCD1 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1 using the numbering given in entry NP_003067.2 (GeneID 6602). | |
100 μL | |
Chromatin Research | |
6602 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction