Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCD1 Mouse anti-Human, Clone: 2F2G6, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 661161IG150UL

Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Two transcript variants encoding different isoforms have been found for this gene.Specifications
SMARCD1 | |
Monoclonal | |
1.05 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96GM5 | |
SMARCD1 | |
SMARCD1 Fusion Protein Ag19100 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
2F2G6 | |
Unconjugated | |
SMARCD1 | |
BAF60A, BRG1 associated factor 60A, CRACD1, Rsc6p, SMARCD1, SWI/SNF complex 60 kDa subunit | |
Mouse | |
Protein G | |
RUO | |
6602 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction