Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552049 Supplier Invitrogen™ Supplier No. PA552049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGPAPGQGLY RSPMPGAAYP RPGMLPGSRM TPQGPSMGPP GYGGNPSVRP GLAQSGMDQS RKRPAPQQIQ QVQQQAVQNR NHNAKK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCD1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMARCD1
Gene Accession No. Q61466, Q96GM5
Gene Alias 60 kDa BRG-1/Brm-associated factor subunit A; AA407987; Baf60a; BRG1-associated factor 60A; chromatin remodeling complex BAF60A subunit; CRACD1; D15Kz1; mammalian chromatin remodeling complex BRG1-associated factor 60A; Protein D15KZ1; Rsc6p; SMARCD1; SWI/SNF complex 60 kDa subunit; SWI/SNF complex 60 kDa subunit A; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1; Swp73-like protein
Gene Symbols SMARCD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMARCD1. Recombinant protein control fragment (Product #RP-102317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363002, 6602, 83797
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.