Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCD1/SMARCD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599831 Supplier Invitrogen™ Supplier No. PA599831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SMARCD1/3.

The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCD1/SMARCD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Smarcd3
Gene Accession No. Q61466, Q6P9Z1, Q6STE5, Q96GM5
Gene Alias 1500001J14Rik; 2210409C08Rik; 60 kDa BRG-1/Brm-associated factor subunit A; 60 kDa BRG-1/Brm-associated factor subunit C; AA407987; Baf60a; Baf60c; BRG1-associated factor 60A; BRG1-associated factor 60C; chromatin remodeling complex BAF60A subunit; chromatin remodeling complex BAF60C subunit; CRACD1; CRACD3; D15Kz1; mammalian chromatin remodeling complex BRG1-associated factor 60A; mammalian chromatin remodeling complex BRG1-associated factor 60C; mBAF60c; Protein D15KZ1; Rsc6p; Smarcd1; SMARCD3; SWI/SNF complex 60 kDa subunit; SWI/SNF complex 60 kDa subunit A; SWI/SNF complex 60 kDa subunit C; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3, isoform 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3; Swp73-like protein
Gene Symbols SMARCD1, Smarcd3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMARCD1(Accession Q96GM5), corresponding to amino acid residues S222-V272.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6602, 6604, 66993, 83797
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.