Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079838
Description
SMARCD2 Polyclonal specifically detects SMARCD2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SMARCD2 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
SMARCD2 | |
The immunogen for this product falls between residue 425 and 475 of human SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 (mammalian chromatin remodeling complex BRG1-associated factor 60B) using the numbering given in entry NP_003068.2 (GeneID 6603). | |
0.1 mL | |
Chromatin Research | |
6603 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation | |
60 kDa BRG-1/Brm-associated factor subunit B, BAF60B, BRG1-associated factor 60B, chromatin remodeling complex BAF60B subunit, CRACD2, mammalian chromatin remodeling complex BRG1-associated factor 60B, PRO2451, Rsc6p, SWI/SNF complex 60 kDa subunit B, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2, Swp73-like protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction