Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541441 Supplier Invitrogen™ Supplier No. PA541441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: STDPQDFIQE WLRSQRRDLK IITDVIGNPE EERRAAFYHQ PWAQEAVGRH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

SMARCD2 is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SMARCD2
Gene Accession No. Q92925
Gene Alias 60 kDa BRG-1/Brm-associated factor subunit B; AW322457; Baf60b; BRG1-associated factor 60B; chromatin remodeling complex BAF60B subunit; CRACD2; mammalian chromatin remodeling complex BRG1-associated factor 60B; PRO2451; Rsc6p; SMARCD2; SWI/SNF complex 60 kDa subunit B; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2; Swp73-like protein
Gene Symbols SMARCD2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SMARCD2 (aa 461-510).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6603
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.