Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCD2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | SMARCD2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SMARCD2 Polyclonal specifically detects SMARCD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SMARCD2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| PBS buffer, 2% sucrose | |
| 6603 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 60 kDa BRG-1/Brm-associated factor subunit B, BAF60B, BRG1-associated factor 60B, chromatin remodeling complex BAF60B subunit, CRACD2, mammalian chromatin remodeling complex BRG1-associated factor 60B, PRO2451, Rsc6p, SWI/SNF complex 60 kDa subunit B, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2, Swp73-like protein | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human SMARCD2 (NP_003068). Peptide sequence STDPQDFIQEWLRSQRRDLKIITDVIGNPEEERRAAFYHQPWAQEAVGRH | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title