Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems



Click to view available options
Analytes:
Ammonia Nitrogen
Copper
Cyanide
Dissolved Oxygen
Iron
Manganese LR
Nitrite
Phosphate HR
Phosphate LR
Silica HR
Sulfate
Zinc HR
pH
Range:
0 to 0.5 ppm
0 to 0.7 ppm
0 to 0.8 ppm
0 to 10 ppm
0 to 100 ppm
0 to 3 ppm
0 to 5 ppm
0 to 6 ppm
0 to 70 ppm
0 to 75 ppm
0.05 to 1 ppm
5 to 9.5 ppm
No. of Tests:
20
25
50
100
Description
Choose from the following test factors:
- Zinc Laboratory Reagent (LR)
- Manganese
- Aluminum
- Ozone
- Cyanide
- Phosphate
- Dissolved Oxygen
- Chromium Hexavalent
- Ammonia Nitrogen LR
Specifications
Specifications
Analytes | Dissolved Oxygen |
For Use With (Application) | Water testing and environmental studies |
For Use With (Equipment) | SMART colorimeter |
No. of Tests | 100 |
Range | 0 to 10 ppm |
Test Method | Winkler Colorimetric |
Type | Dissolved Oxygen Reagent System |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction