Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SMART Digest™ Trypsin Kits

Catalog No. 03251711
Change view
Click to view available options
Format:
Bulk Low pH Magnetic Resin
Bulk Low pH Resin
Bulk Magnetic Resin
Bulk Non-magnetic Resin
Resin in Pre-filled PCR Tubes
Soluble Trypsin
Type:
Soluble Trypsin Kit
Soluble Trypsin Kit - Low Salt
Soluble Trypsin Kit - Low pH
Trypsin Kit
Includes:
Low pH Resin, Collection Plate
Resin, Buffer
Resin, Buffer, Collection Plate, Filter Plate
Resin, Buffer, Collection Plate, SOLAμ SPE Plate
Resin, Collection Plate
Resin, Collection Plate, Filter Plate
Resin, Collection Plate, SOLAμ SPE Plate
Soluble Trypsin, Buffer, Collection Plate
Soluble Trypsin, Low pH Buffer, Collection Plate
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Type Includes
03-251-711 Soluble Trypsin Soluble Trypsin Kit Soluble Trypsin, Buffer, Collection Plate
03-150-404 Resin in Pre-filled PCR Tubes Trypsin Kit Resin, Collection Plate
03-251-893 Bulk Low pH Resin Trypsin Kit Resin, Collection Plate
03-251-897 Resin in Pre-filled PCR Tubes Trypsin Kit Low pH Resin, Collection Plate
03-150-406 Resin in Pre-filled PCR Tubes Trypsin Kit Resin, Collection Plate, SOLAμ SPE Plate
03-251-894 Bulk Low pH Magnetic Resin Trypsin Kit Resin, Collection Plate
03-150-405 Resin in Pre-filled PCR Tubes Trypsin Kit Resin, Collection Plate, Filter Plate
03-375-364 Bulk Non-magnetic Resin Trypsin Kit Resin, Buffer
03-253-935 Soluble Trypsin Soluble Trypsin Kit - Low pH Soluble Trypsin, Low pH Buffer, Collection Plate
03-252-203 Bulk Magnetic Resin Trypsin Kit Resin, Buffer
03-252-204 Bulk Non-magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, Filter Plate
13-622-452 Bulk Magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, Filter Plate
03-252-205 Bulk Non-magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, SOLAμ SPE Plate
03-252-206 Bulk Magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, SOLAμ SPE Plate
03-170-489 Soluble Trypsin Soluble Trypsin Kit - Low Salt Soluble Trypsin, Buffer, Collection Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 03-251-711 Supplier Thermo Scientific™ Supplier No. 60113101
Add to Cart
Add to Cart

Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.

Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

Trypsin Protein Digestion

SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.

Easy and simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.

Couple with SOLAμ SPE Plates for Desalting

When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

TRUSTED_SUSTAINABILITY

Specifications

Type Soluble Trypsin Kit
Description SMART Digest Trypsin Kit, Soluble
Format Soluble Trypsin
Enzyme Trypsin
Cleavage C-terminal side of lysine and arginine amino acids
Capture None
Product Line SMART Digest
For Use With (Application) Peptide Mapping
Includes Soluble Trypsin, Buffer, Collection Plate
Can I use the SMART Digest Soluble Trypsin Kit with filter-aided sample preparation (FASP)?

Yes, we recommend the following protocol, which can be optimized as needed:
1. Rinse a Fisher Scientific Vivacon 500, 30kDa MWCO filter (Cat. No. 14558347) to remove trace amounts of glycerin using a fill volume of water. Decant the filtrate and collection vessel. The filter is now ready for use. If you do not want to use the Vivacon filter immediately, then store the pre-rinsed device in the refrigerator with water or buffer covering the membrane surface. Do not allow the membrane to dry out.
2. Dilute sample water to a total volume of 150 µL on the filter.
3. Centrifuge 14,000 RPM for 15 minutes and discard the flow-through.
4. Add 150 µL water.
5. Centrifuge 14,000 RPM for 15 minutes and discard the flow-through.
6. Add 200 µL of SMART Digest buffer.
7. Change the collection vial and add 5 µL of the SMART Digest soluble enzyme to the filter.
8. Parafilm the filters to prevent evaporation of sample during digestion. Incubate the units in a heated bath at 70°C for 60 - 90 minutes.
9. Optional: As needed, add DTT (3 µL of 50 mM DTT), heat at 37°C and incubate for 30 minutes. Alkylate as needed (using 3 µL of 150 mM alkylating reagent).
10. After incubation, add 2 µL of 1% trifluoroacetic acid to the filter. Mix for 1 minute.
11. Centrifuge the filter units at 14,000 RPM for 10 min.
12. As needed, measure protein concentration using a Thermo Scientific NanoDrop Spectrophotometer, or similar procedure.

What is the best way to automate my workflow with the SMART Digest Soluble Trypsin Kit?

To automate the SMART Digest Soluble Trypsin Kit workflow, we recommend using deepwell plates with Sepra Seals, or PCR plates with iron-on seals in combination with an automated liquid handling platform. Any heating unit with a heated lid, capable of providing uniform heating at 70°C may be used.

Can I use chaotropes with the SMART Digest Soluble Trypsin Kit?

The kit is compatible with up to 1M urea, but incompatible with most concentrations of guanidine.

Can I use surfactants with the SMART Digest Soluble Trypsin Kit?

The kit is compatible with many surfactants including OGS, CHAPS, Nonidet, and NP-40.

Does digestion at high temperatures using the SMART Digest Soluble Trypsin Kit result in an increase in chemical modifications?

Thus far, studies have shown that by accelerating the reaction and minimizing the reaction time, fewer chemical modifications are observed in comparison to protocols utilizing elongated digestion times at lower temperatures. For more details, refer to the application note titled 'SMART Digest Compared to Classic In-Solution Digestion of Rituximab for In-Depth Peptide Mapping Characterization'.

Will disulfide bonds scramble during digestion with the SMART Digest Soluble Trypsin Kit?

See the publication, 'Thiol-Disulfide Exchange in Human Growth Hormone' for details (Pharm Res (2016) 33:1370–1382 DOI 10.1007/s11095-016-1879-3). No disulfide bond rearrangement was observed. However, free cysteines could potentially lead to scrambling. Alkylate free cysteines to prevent disulfide bond scrambling, as needed.

Do I have to reduce and alkylate my protein when using the SMART Digest Soluble Trypsin Kit?

The SMART Digest Soluble Trypsin Kit was engineered to be thermally stable. When operated at high temperatures (e.g. 70°C), denaturation and digestion happen simultaneously. Therefore, for many quantitative workflows there is no need to perform the additional steps of denaturation, reduction and alkylation. However, during this process many disulfide bonds will remain intact. As such, for many identification workflows it is recommended that you perform reduction and alkylation after digestion. In our experience, denaturants and reducing reagents negatively impact digestion when using the SMART Digest Soluble Trypsin Kits.

What is the pH of the SMART Digest buffer that comes in the SMART Digest Soluble Trypsin Kit?

The pH of the digestion buffer as received in the SMART Digest Soluble Trypsin Kit is approximately 8. When used at 70°C, the pH of the buffer is approximately 7. Operation at this lower pH helps to prevent the formation of artificial modifications.

Are there salts in the SMART Digest buffer that comes in the SMART Digest Soluble Trypsin Kit?

The SMART Digest buffer in the SMART Digest Soluble Trypsin Kit contains ~ 0.5 M salts. The salts greatly aid in achieving rapid digestion at high temperatures. Desalting through the use of valve switching is highly advised, although the use of SPE cleanup has also been successful.

Do I have to use the SMART Digest buffer that comes in the SMART Digest Soluble Trypsin Kit?

The SMART Digest buffer was optimized for maximum trypsin activity at elevated temperatures. Other buffers can be used, but their use may negatively impact trypsin activity. If your application requires the use of an alternative buffer, digestion time and temperature should be optimized accordingly.

How long should I digest my sample when using the SMART Digest Soluble Trypsin Kit?

All proteins vary with regards to digestion so you will need to adjust the temperature and incubation time accordingly. We recommend the following strategy for screening digestion time:
1. Create a method in your heating unit setting the desired operating temperature (we recommend 70°C). Start this method and allow the temperature to reach equilibrium for at least 5 minutes before adding samples.
2. Prepare eight identical samples using a relatively high known concentration of native analyte in the matrix of operation, diluting them to 50 µL each with ultrapure water, if necessary.
3. Add each sample along with 150 µL of SMART Digest buffer and 5 µL of SMART Digest soluble enzyme to each of the eight wells.
4. Place all samples firmly into the preheated unit.
5. Periodically (e.g. every 15 minutes) remove a sample and quench the reaction using an equivalent volume of 1% trifluoroacetic acid, or 1% formic acid.
6. Following incubation analyze the samples to determine the extent of digestion.
7. Once the intact protein peak has disappeared, digestion is complete and the corresponding digestion time can be used for subsequent analyses.

Do not reduce and alkylate sample prior to digestion. Our findings indicate that the chemicals used for denaturation, reduction and alkylation may negatively impact the activity of the enzyme and the solubility of the protein sample. If the peptides of interest require reduction prior to analysis it is recommended that these steps be performed post digestion.

Do I need to vary the amount of trypsin used depending on protein load when I am using the SMART Digest Soluble Trypsin Kit?

Generally, there is no requirement to vary the amount of trypsin used for any given sample. Studies have shown that when operating at elevated temperatures with a stable enzyme, the reactions are significantly less concentration dependent than traditional protocols. For samples containing less than 5 µg of total protein, titering the enzyme amount may prevent an excess of enzyme from interfering with the analysis.

How much material can I digest with the SMART Digest Soluble Trypsin Kit?

For every 5 µL of the SMART Digest Soluble Trypsin Kit up to 50 µL of plasma (approx. 3.5 mg of protein) is readily digested.

When I use 5 microliters of the SMART Digest Soluble Trypsin Kit, how much trypsin is present?

The trypsin enzyme concentration is 1 µg/µL so there would be 5 µg of trypsin in 5µL.

Do I need to place my sample on a shaker when using the SMART Digest Soluble Trypsin Kit?

As long as the substrate is soluble, shaking is not required.

What equipment do I need to use a SMART Digest Soluble Trypsin Kit?

We recommend using any heating unit with a heated lid that is capable of providing uniform heating at 70°C.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.