Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ SMART™ Spectro System COD Reagents



Click to view available options
Range:
0 to 15,000ppm
0 to 1500ppm
0 to 150ppm
Description
Measure low, medium or high levels of Chemical Oxygen Demand (COD) using a mercury reagent system approved by the Environmental Protection Agency (EPA).
- Comes in 25 vials to a case
- Requires COD Heater Block, not included
Specifications
Specifications
Certifications/Compliance | EPA |
For Use With (Equipment) | LaMotte™ colorimeters |
Range | 0 to 15,000ppm |
Test Method | Acid digestion |
Length (English) | 0.62 in. |
Length (Metric) | 1.6 cm |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction