Learn More
Thermo Scientific™ Smart-Vue Pro Accessories
Customize your Smart-Vue Pro remote monitoring system with accessories to meet your specific needs.
Supplier:  Thermo Scientific™ SVPACCSRC001
 View more versions of this product
Catalog No. SVACCSRC001
Only null left
Add to Cart
Description
Description
Choose from an assorment of Smart-Vue Pro accessories including power supplies, cables and mounting hardware, antenna and audible alarms to customize your remote monitoring system as well as ensure system integrity.
Compatible with Smart-Vue Pro remote monitoring system.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Smart-Vue Pro Dry contact alert device with LoRaWANTM wireless connectivity 915 Mhz for use with LoRaWANTM systems
Smart-Vue Pro Remote Monitoring System
915 MHz
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.