Learn More
Thermo Scientific™ Smart-Vue Pro Alert Software
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
$205.00 - $79150.00
Specifications
Type | Smart-Vue Pro Software and Subscription Service |
---|---|
For Use With (Equipment) | Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
SVSWSA20005
|
Thermo Scientific™
SVPSWSA20005 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 5 PTS |
Each for $205.00
|
|
|||||
SVSWSA20020
|
Thermo Scientific™
SVPSWSA20020 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 20 PTS |
Each for $816.50
|
|
|||||
SVSWSA20050
|
Thermo Scientific™
SVPSWSA20050 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 50 PTS |
Each for $2,041.09
|
|
|||||
SVSWSA20100
|
Thermo Scientific™
SVPSWSA20100 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 100 PTS |
Each for $4,050.00
|
|
|||||
SVSWSA20250
|
Thermo Scientific™
SVPSWSA20250 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 250 PTS |
Each for $10,021.07
|
|
|||||
SVSWSA20500
|
Thermo Scientific™
SVPSWSA20500 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 500 PTS |
Each for $20,211.05
|
|
|||||
SVSWSA21000
|
Thermo Scientific™
SVPSWSA21000 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 1000 PTS |
Each for $40,160.94
|
|
|||||
SVSWSA21500
|
Thermo Scientific™
SVPSWSA21500 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 1500 PTS |
Each for $60,000.00
|
|
|||||
SVSWSA22000
|
Thermo Scientific™
SVPSWSA22000 |
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 2500 PTS |
Each for $79,150.00
|
|
|||||
Description
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions, Smart-Vue™ Alert will automatically notify the user based notification preferences wherever you are (phone, text or email).
- Add-on notification package through SMS and voice calls (Twillo)
- 12 month license (post-activation)
- License depends on the number of monitoring points desired
Specifications
Smart-Vue Pro Software and Subscription Service | |
Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.