Learn More
Thermo Scientific™ Smart-Vue Pro Modules
Increase efficiency with multi-sensor wireless monitoring
Supplier:  Thermo Scientific™ SVPHWRMOD004
 View more versions of this product
Catalog No. SVHWRMOD004
Item Discontinued This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.
Description
Description
Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment
Bright 2.4 in. color touch screen with multiple views Alarms locally with real -time audio-visual alarms on the modules Audio visual alarms for local notification Communication locally via Bluetooth and to the system via LoRaLow AC power consumption through USB port access Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring Choice of wired and wireless sensors with automatic sensor detection User traceability with customized levels of user access with encrypted password protection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Module
Not Calibrated
3.6 V Lithium
Smart-Vue Pro Smart-Vue Pro Quattro with multifrequencies support. Delivered with two 3.6 V lithium batteries, two sensor mounting kit, one mounting holder, one antena and two 1.5 m/5 ft. flat cable.
2 to 4
No Display
Audible
3.6
Two 3.6 V Lithium Batteries, Two Sensor Mounting Kits, One Mounting Holder, One Antenna, and Two 1.5 m/5 ft. Flat Cables
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.