Learn More
Thermo Scientific™ Smart-Vue Pro Receivers
Catalog No. 11731060
Change view
Click to view available options
Includes:
Smart-Vue Pro Indoor Receiver for LoRa Modules (Enable Ethernet or WiFi Connectivity) 915 Mhz with Universal Power Supply, External Antenna, Universal Power Supply, LoRaWAN Flyer
Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with Antenna, Power Supply, 915 Mhz, no data/SIM
Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with SIM card, data plan for up to 10 devices/1 year. Includes Antenna, Power Supply, 915 Mhz
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
11-731-060 Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with SIM card, data plan for up to 10 devices/1 year. Includes Antenna, Power Supply, 915 Mhz
11-731-063 Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with Antenna, Power Supply, 915 Mhz, no data/SIM
SVHWRLGW001 Smart-Vue Pro Indoor Receiver for LoRa Modules (Enable Ethernet or WiFi Connectivity) 915 Mhz with Universal Power Supply, External Antenna, Universal Power Supply, LoRaWAN Flyer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-731-060 Supplier Thermo Scientific™ Supplier No. SVPHWAPG915C
Add to Cart
Add to Cart
IT friendly LoRaWan provides easy set-up of a reliable, independent and secure wireless network
LoRA is short for Long Range wireless Wide Area Network. It is a stand-alone dedicated network and increases connectivity range for IoT devices by 2 km in urban and 10 km in rural areas. This receiver pushes data to the Cloud by connecting to your local network via an Ethernet or Wi-Fi connection.
Hardware
Receivers
Multitech Advanced receivers (blue) support up to 1000 data logging sensors. Gateways are available in 915 MHz for North America and 865-868 MHz for rest of world.
Multitech Pro receivers (white) Support for up to 50 data logging sensors per gateway and are available in Ethernet and 4G options in 915 MHz for North America and 865-868 MHz for rest of world.
Modules
Smart-Vue Pro modules collect data from independent sensors connected to equipment from any OEM. Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment.
Sensors
The Smart-Vue Pro sensors collect data points to be logged from any laboratory equipment OEM. Sensor kits, including gateways are available.
Subscription
Requires subscription
Availability
Check with your specialist for country availability. Receivers are available in 865-868 MHz and 915 MHz.
TRUSTED_SUSTAINABILITY
Specifications
Type Remote Monitoring Gateway
Display Type No Display
Data Memory NA
Calibration NA
Alarm Types E-mail, SMS, Audible, Telephone, Mobile Push Notification
Includes Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with SIM card, data plan for up to 10 devices/1 year. Includes Antenna, Power Supply, 915 Mhz
IP Rating IP66/68
Model PRO MTC Receiver for up to 50 Modules
Quantity 1
Warranty 1 Year Warranty
For Use With (Equipment) Smart-Vue Pro Remote Monitoring