Learn More
Thermo Scientific™ Smart-Vue Pro On-Premise Software and Subscription Services
A simplified software experience, allowing you to focus on what matters the most
$662.00 - $228900.00
Specifications
Type | Smart-Vue Pro Software and Subscription Service |
---|---|
Certifications/Compliance | Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
SVSWPS20005
|
Thermo Scientific™
SVPSWPS20005 |
Smart-Vue Pro Web application on-premise license up to 5 measurement points |
Each for $1,165.79
|
|
|||||
SVSWPS20020
|
Thermo Scientific™
SVPSWPS20020 |
Smart-Vue Pro Web application on-premise license up to 20 measurement points |
Each for $4,680.86
|
|
|||||
SVSWPS20050
|
Thermo Scientific™
SVPSWPS20050 |
Smart-Vue Pro Web application on-premise license up to 50 measurement points |
Each for $5,830.82
|
|
|||||
SVSWPS20100
|
Thermo Scientific™
SVPSWPS20100 |
Smart-Vue Pro Web application on-premise license up to 100 measurement points |
Each for $12,297.88
|
|
|||||
SVSWPS20250
|
Thermo Scientific™
SVPSWPS20250 |
Smart-Vue Pro Web application on-premise license up to 250 measurement points |
Each for $29,208.18
|
|
|||||
SVSWPS20500
|
Thermo Scientific™
SVPSWPS20500 |
Smart-Vue Pro Web application on-premise license up to 500 measurement points |
Each for $59,257.94
|
|
|||||
SVSWPS21000
|
Thermo Scientific™
SVPSWPS21000 |
Smart-Vue Pro Web application on-premise license up to 1000 measurement points |
Each for $115,050.00
|
|
|||||
SVSWPS21500
|
Thermo Scientific™
SVPSWPS21500 |
Smart-Vue Pro Web application on-premise license up to 1500 measurement points |
Each for $171,557.42
|
|
|||||
SVSWPS22000
|
Thermo Scientific™
SVPSWPS22000 |
Smart-Vue Pro Web application on-premise license up to 2000 measurement points |
Each for $228,900.00
|
|
|||||
SVSWPU20020
|
Thermo Scientific™
SVPSWPU20020 |
Smart-Vue Pro Web application on-premise license upgrade - from 5 to 20 measurement points |
Each for $3,520.87
|
|
|||||
SVSWPU20050
|
Thermo Scientific™
SVPSWPU20050 |
Smart-Vue Pro Web application on-premise license upgrade - from 20 to 50 measurement points |
Each for $4,480.00
|
|
|||||
SVSWPU20100
|
Thermo Scientific™
SVPSWPU20100 |
Smart-Vue Pro Web application on-premise license upgrade - from 50 to 100 measurement points |
Each for $5,830.82
|
|
|||||
SVSWPU20250
|
Thermo Scientific™
SVPSWPU20250 |
Smart-Vue Pro Web application on-premise license upgrade - from 100 to 250 measurement points |
Each for $17,427.58
|
|
|||||
SVSWPU20500
|
Thermo Scientific™
SVPSWPU20500 |
Smart-Vue Pro Web application on-premise license upgrade - from 250 to 500 measurement points |
Each for $29,107.81
|
|
|||||
SVSWPU21000
|
Thermo Scientific™
SVPSWPU21000 |
Smart-Vue Pro Web application on-premise license upgrade - from 500 to 1000 measurement points |
Each for $58,057.71
|
|
|||||
SVSWPU21500
|
Thermo Scientific™
SVPSWPU21500 |
Smart-Vue Pro Web application on-premise license upgrade - from 1000 to 1500 measurement points |
Each for $86,650.00
|
|
|||||
SVSWPU22000
|
Thermo Scientific™
SVPSWPU22000 |
Smart-Vue Pro Web application on-premise license upgrade - from 1500 to 2000 measurement points |
Each for $115,600.00
|
|
|||||
SVSWSM20005
|
Thermo Scientific™
SVPSWSM20005 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 5 PTS |
Each for $662.00
|
|
|||||
SVSWSM20020
|
Thermo Scientific™
SVPSWSM20020 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 20 PTS |
Each for $2,670.00
|
|
|||||
SVSWSM20050
|
Thermo Scientific™
SVPSWSM20050 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 50 PTS |
Each for $3,320.18
|
|
|||||
SVSWSM20100
|
Thermo Scientific™
SVPSWSM20100 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 100 PTS |
Each for $6,620.00
|
|
|||||
SVSWSM20250
|
Thermo Scientific™
SVPSWSM20250 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 250 PTS |
Each for $16,591.79
|
|
|||||
SVSWSM20500
|
Thermo Scientific™
SVPSWSM20500 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 500 PTS |
Each for $33,201.85
|
|
|||||
SVSWSM21000
|
Thermo Scientific™
SVPSWSM21000 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 1000 PTS |
Each for $66,200.00
|
|
|||||
SVSWSM21500
|
Thermo Scientific™
SVPSWSM21500 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 1500 PTS |
Each for $94,850.00
|
|
|||||
SVSWSM22000
|
Thermo Scientific™
SVPSWSM22000 |
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 2000 PTS |
Each for $114,450.00
|
|
|||||
Description
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times. The on-premise solution maintains your Smart-Vue Pro data locally on your site's server.
- Intuitive dashboard simplifies daily usage.
- Optional floor plan and sites views.
- Data can be exported with a click for reporting and customizable graphs.
- Web application on Cloud or customer server.
- Instant access to alarms, readings, and graphs.
- Remote data access.
- Flexible organization by site, department user, equipment, and criticality.
- Audit trail and secure-level access control to support 21CFR Part 11
Specifications
Smart-Vue Pro Software and Subscription Service | |
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.