Learn More
Thermo Scientific™ Smart-Vue™ Receivers
Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.
Supplier: Thermo Scientific™ SV103509LSB
Description
Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifications
Receiver, 915MHz | |
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. | |
915 MHz |
USB 1.0 or 2.0 Receiver, 25mW+SW 99 sensors | |
Wireless Monitoring Solution |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.