Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC1-alpha Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993107
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3107 10 μL
50-199-3106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3107 Supplier Active Motif Supplier No. 61068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Structural Maintenance of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregatior defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.

Specifications

Antigen SMC1-alpha
Applications ChIP sequencing (ChIP-seq), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene SMC1B
Gene Accession No. Q14683, Q8NDV3
Gene Alias 14S cohesin SMC1 subunit; 5830426I24Rik; BOS_26006; BOS_6225; CDLS2; Chromosome segregation protein SmcB; DKFZp686L19178; DXhXs423e; DXS423E; KIAA0178; MGC138332; mitosis-specific chromosome segregation protein like protein beta; mKIAA0178; RP6-29D12.1; SB1.8; segregation of mitotic chromosomes 1; segregation of mitotic chromosomes b; segregation of mitotic chromosomes-like 1; SM1A; SMC; SMC (segregation of mitotic chromosomes 1)-like 1; SMC protein; SMC protein 1A; SMC protein 1B; SMC1; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; SMC1 protein cohesin subunit; SMC1 structural maintenance of chromosomes 1-like 1; SMC1 structural maintenance of chromosomes 1-like 2; Smc1a; SMC-1A; smc1a.L; smc1aa; SMC1alpha; SMC-1-alpha; SMC1alpha protein; Smc1b; SMC-1B; SMC1BETA; SMC-1-beta; SMC1L1; SMC1L2; Smcb; SMC-like 1; SMC-protein; structural maintenace of chromosomes 1B; structural maintenance of chromosomes 1; structural maintenance of chromosomes 1 like 1; structural maintenan
Gene Symbols SMC1A, SMC1B
Host Species Rabbit
Immunogen This SMC-alpha antibody was raised against a peptide within the C-terminal region of human SMC1-alpha.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27127, 8243
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.