Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMC1, p Ser966 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
Supplier: Novus Biologicals NB100206
Description
SMC1 Polyclonal specifically detects SMC1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
SMC1 (p Ser966) | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, Sb1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, White-tufted-ear marmoset, Crab-eating macaque, Small-eared galago and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:500 - 1:5000, Immunoprecipitation 2 - 5 ug/mg of lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000 | |
Q14683 | |
SMC1A | |
Immunogen for this antibody was a phosphorylated synthetic peptide, which represented a portion of human Structural Maintenance of Chromosomes 1 (GeneID 8243) around serine 966 according to the numbering given in entry NP_006297.2. | |
100 μL | |
Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Phospho Specific | |
8243 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction