Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMC1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588855 Supplier Invitrogen™ Supplier No. PA588855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILLKTAAPNL RALENLKTVR DKFQESTDAF EASRKEARLC RQEFEQVKKR RYDLFTQCFE HVSISIDQIY KKLCRNNSAQ VSSYIKEQTQ DQFQMIVISL KEEFYSRADA LIGIYPEYDD CMFSRVLTLD LSQYPDTEGQ ESSKRHGESR; Positive Samples: Rat testis; Cellular Location: Chromosome, Nucleus, centromere.

The SMC1B gene encodes a meiosis-specific subunit of the cohesin complex, which is essential for proper chromosome segregation during meiosis. It plays a critical role in sister chromatid cohesion and DNA recombination. The SMC1B protein, also known as SMC1 beta, forms a heterodimer with SMC3, which is necessary for the formation of the cohesin complex that holds sister chromatids together until anaphase. Studies have demonstrated that SMC1B is required for the activation of the spindle assembly checkpoint (SAC) during oocyte meiosis. The loss of SMC1B function leads to improper chromosome alignment and segregation, resulting in infertility due to meiotic arrest. Furthermore, the gene is expressed exclusively during meiosis, and mutations or deletions within SMC1B can cause sterility due to disruptions in meiotic progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Smc1b
Gene Alias bK268H5; bK268H5.5; mitosis-specific chromosome segregation protein like protein beta; RP1-102D24.3; SMC; SMC protein 1B; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; SMC1 structural maintenance of chromosomes 1-like 2; SMC1B; SMC-1B; SMC1BETA; SMC-1-beta; SMC1L2; structural maintenace of chromosomes 1B; structural maintenance of chromosomes 1B; structural maintenance of chromosomes protein 1B
Gene Symbols Smc1b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1012-1161 of human SMC1B (NP_0012784301).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300121
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.