Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMC1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522199
Description
Recommended positive controls: A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
SMC1B | |
Polyclonal | |
Unconjugated | |
Smc1b | |
bK268H5; bK268H5.5; mitosis-specific chromosome segregation protein like protein beta; RP1-102D24.3; SMC; SMC protein 1B; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; SMC1 structural maintenance of chromosomes 1-like 2; SMC1B; SMC-1B; SMC1BETA; SMC-1-beta; SMC1L2; structural maintenace of chromosomes 1B; structural maintenance of chromosomes 1B; structural maintenance of chromosomes protein 1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M Tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8NDV3 | |
Smc1b | |
Recombinant fragment corresponding to a region within amino acids 154 and 433 of SMC1B (Uniprot ID#Q8NDV3). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction