Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578335
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (90%), Rat (92%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAP-E and CAP-C (Chromosome Associated Protein-E & C) are also SMC (Structural Maintenance of Chromosome) family members that form a heterodimeric complex required for mitotic chromosome condensation to achieve proper segregation of genetic information during subsequent cell division. hCAP-C/hCAP-E is a component of a multiprotein complex called condensin that interacts with phosphorylation of Histone H3 resulting in the mitotic chromosome condensation. The distribution patterns of the two heterodimeric complexes in interphase nucleus indicate independent behavior of the two complexes during cell cycle. These results suggest that the two distinct complexes are involved in different aspects of mitotic chromosome organization in humar cells.
Specifications
SMC2 | |
Polyclonal | |
Unconjugated | |
SMC2 | |
5730502P04Rik; AI255214; AW545314; CAP E; CAPE; CAP-E; Chromosome-associated protein E; FGF-inducible protein 16; fibroblast growth factor inducible 16; Fin16; FLJ10093; hCAP-E; PRO0324; SMC protein 2; Smc2; SMC-2; SMC2 (structural maintenance of chromosomes 2, yeast)-like 1; SMC2 protein; SMC2 structural maintenance of chromosomes 2-like 1; Smc2l1; structural maintenance of chromosomes (SMC) family member, chromosome-associated protein E; structural maintenance of chromosomes 2; structural maintenance of chromosomes protein 2; XCAP-E homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10592 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O95347 | |
SMC2 | |
Recombinant protein encompassing a sequence within the center region of human SMC2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction