Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC3, Mouse anti-Human, Clone: 1G1, Abnova™

Catalog No. 50858364 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-858-364 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-858-364 Supplier Abnova Corporation Supplier No. H00009126M01

Mouse Monoclonal Antibody

Sequence: IDQLMNQMQQIETQQRKFKASRDSILSEMKMLKEKRQQSEKTFMPKQRSLQSLEASLHAMESTRESLKAELGTDLLSQLSLEDQKRVDALNDEIRQLQQE

Specifications

Antigen SMC3
Applications ELISA
Classification Monoclonal
Clone 1G1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene structural maintenance of chromosomes 3
Gene Accession No. BC047324
Gene Alias BAM/BMH/CDLS3/CSPG6/HCAP/SMC3L1
Gene Symbols SMC3
Host Species Mouse
Immunogen CSPG6 (AAH47324, 711 a.a. ∼ 810 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9126
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.