Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMC3, p Ser383 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100208AF647
Description
SMC3 Polyclonal antibody specifically detects SMC3 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SMC3 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Bamacan, BAMSMC protein 3, Basement membrane-associated chondroitin proteoglycan, BMH, Chondroitin sulfate proteoglycan 6, chondroitin sulfate proteoglycan 6 (bamacan), Chromosome-associated polypeptide, CSPG6structural maintenance of chromosomes protein 3, hCAP, HCAPbamacan, SMC-3, SMC3L1CDLS3, structural maintenance of chromosomes 3 | |
Immunogen for this antibody was a phosphorylated synthetic peptide, which represented a portion of human Structural Maintenance of Chromosomes 3 (GeneID 9126) around serine 383 according to the numbering given in entry NP_005436.1. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Phospho Specific | |
9126 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction