Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMC3 Recombinant Rabbit Monoclonal Antibody (BLR099G)
GREENER_CHOICE

Catalog No. PIMA544389
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544389 100 μL
PIMA544390 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544389 Supplier Invitrogen™ Supplier No. MA544389
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SMC3 Recombinant Monoclonal Antibody for Western Blot, Flow, IP

This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintenance of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC3
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BLR099G
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.09% sodium azide; pH 7.8-8.6
Gene SMC3
Gene Accession No. Q9UQE7
Gene Alias Bam; Bamacan; basement membrane chondroitin sulfate proteoglycan; basement membrane-associated chondroitin proteoglycan; BMH; CDLS3; chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); chromosome segregation protein SmcD; Chromosome-associated polypeptide; cspg6; HCAP; im:7142991; mad member-interacting protein 1; Mmip1; SMC protein 3; Smc3; SMC-3; SMC3 protein; SMC3L1; SmcD; structural maintenace of chromosomes 3; structural maintenance of chromosomes 3; structural maintenance of chromosomes protein 3; structural maintenance of chromosomes protein 3-like protein; wu:fb22e01; wu:fc30d07
Gene Symbols SMC3
Host Species Rabbit
Immunogen Peptide representing a region between residue 1175 and the C-terminus (residue 1217) of human Structural maintenance of chromosomes protein 3 (SMC3).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9126
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.