Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMC4 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100375AF647
Description
SMC4 Polyclonal antibody specifically detects SMC4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SMC4 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog | |
A synthetic peptide, which represented a portion of human structural maintenance of chromosomes 4 encoded within exon 23 (LocusLink ID 10051). According to Swiss-Prot entry Q9NTJ3, the peptide is near the C-terminus. | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
10051 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction