Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMC4 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100374G
Description
SMC4 Polyclonal antibody specifically detects SMC4 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SMC4 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog | |
The immunogen recognized by this antibody maps to a region between residues 600 and 650 of human Structural Maintenance of Chromosomes 4 using the numbering given in entry NP_005487.2 (GeneID 10051). | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
10051 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction