Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556150
Description
Immunogen sequence: KLTQEETNFK SLVHDLFQKV EEAKSSLAMN RSRGKVLDAI IQEKKSGRIP GIYGRLGDLG AIDEKYDVAI SSCCHALDYI VVDSIDIAQE CVNFLKRQNI GVATFIGLDK MAVWAKKMTE IQTPENTPRL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.
Specifications
SMC4 | |
Polyclonal | |
Unconjugated | |
Smc4 | |
2500002A22Rik; C79747; Capc; CAP-C; chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4; SMC-4; SMC4 structural maintenance of chromosomes 4-like 1; SMC4L1; structural maintenance of chromosomes 4; structural maintenance of chromosomes protein 4; XCAP-C homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10051, 295107, 70099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8CG47, Q9NTJ3 | |
Smc4 | |
Recombinant protein corresponding to Human SMC4. Recombinant protein control fragment (Product #RP-92412). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction