Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987963
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7963 Supplier Bioss Supplier No. BS7728R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.

Specifications

Antigen SMC4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Smc4
Gene Accession No. Q9NTJ3
Gene Alias 2500002A22Rik; C79747; Capc; CAP-C; Chromosome-associated polypeptide C; hCAP-C; SMC protein 4; Smc4; SMC-4; SMC4 structural maintenance of chromosomes 4-like 1; SMC4L1; structural maintenance of chromosomes 4; Structural maintenance of chromosomes protein 4; XCAP-C homolog
Gene Symbols Smc4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SMC4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10051
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.