Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598595
Description
SMC5 Polyclonal Antibody for Western Blot, ELISA
Core component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs). Required for recruitment of telomeres to PML nuclear bodies. Required for sister chromatid cohesion during prometaphase and mitotic progression; the function seems to be independent of SMC6. SMC5-SMC6 complex may prevent transcription of episomal DNA, such as circular viral DNA genome.
Specifications
SMC5 | |
Polyclonal | |
Unconjugated | |
Smc5 | |
hSMC5; KIAA0594; mKIAA0594; MLZ-453; mSMC5; Protein expressed in male leptotene and zygotene spermatocytes 453; SMC protein 5; Smc5; SMC-5; SMC5 structural maintenance of chromosomes 5-like 1; Smc5l1; structural maintenance of chromosomes 5; structural maintenance of chromosomes protein 5 | |
Rabbit | |
Protein G | |
RUO | |
23137 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8IY18 | |
Smc5 | |
Recombinant Human Structural maintenance of chromosomes protein 5 protein (296-403AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction