Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115931 Supplier Thermo Scientific Supplier No. PA5115931
Add to Cart
Add to Cart

SMC5 Polyclonal Antibody

SMC5 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Smc5
Gene Accession No. Q8CG46, Q8IY18
Gene Alias hSMC5; KIAA0594; mKIAA0594; MLZ-453; mSMC5; Protein expressed in male leptotene and zygotene spermatocytes 453; SMC protein 5; Smc5; SMC-5; SMC5 structural maintenance of chromosomes 5-like 1; Smc5l1; structural maintenance of chromosomes 5; structural maintenance of chromosomes protein 5
Gene Symbols Smc5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMC5(Accession Q8IY18), corresponding to amino acid residues E964-P1014.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226026, 23137, 293967
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.