Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560096 Supplier Invitrogen™ Supplier No. PA560096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEEHGKIKRE ELDVKHALSY NQRQLKELKD SKTDRLKRFG PNVPALLEAI DDAYRQGHFT YKPVGPLGAC IHLRDPELAL AIESCL Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 84%.

Core component of the SMC5-SMC6 complex, a complex involved in DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs). Required for recruitment of telomeres to PML nuclear bodies. SMC5-SMC6 complex may prevent transcription of episomal DNA, such as circular viral DNA genome (PubMed:26983541). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMC6
Gene Accession No. Q96SB8
Gene Alias 2810489L22Rik; 3830418C19Rik; AA990493; AU018782; AW742439; hSMC6; Kiaa4103; mKIAA4103; mSMC6; SMC protein 6; Smc6; SMC-6; SMC6 structural maintenance of chromosomes 6-like 1; SMC6L1; structural maintenance of chromosomes 6; structural maintenance of chromosomes protein 6; Structural maintenance of chromosomes protein 6-like protein
Gene Symbols SMC6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMC6. Recombinant protein control fragment (Product #RP-96173).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79677
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.