Learn More
SMC6 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300237A


Description
Immunohistochemistry: Epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
SMC6 Polyclonal antibody specifically detects SMC6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
SMC6 | |
Polyclonal | |
Unconjugated | |
SMC6 | |
2810489L22Rik; 3830418C19Rik; AA990493; AU018782; AW742439; hSMC6; Kiaa4103; mKIAA4103; mSMC6; SMC protein 6; Smc6; SMC-6; SMC6 structural maintenance of chromosomes 6-like 1; SMC6L1; structural maintenance of chromosomes 6; structural maintenance of chromosomes protein 6; Structural maintenance of chromosomes protein 6-like protein | |
Antigen affinity chromatography | |
RUO | |
67241, 79677 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
phosphate, tris citrate with 0.09% sodium azide | |
Q924W5, Q96SB8 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.