Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987965
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7965 Supplier Bioss Supplier No. BS7730R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Members of the parvin family, including PARVA, are actin-binding proteins associated with focal contacts.

Specifications

Antigen SMC6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SMC6
Gene Accession No. Q96SB8
Gene Alias 2810489L22Rik; 3830418C19Rik; AA990493; AU018782; AW742439; DNA repair protein RHC18; DNA repair protein SMC6; hSMC6; Kiaa4103; L3502.2; mKIAA4103; mSMC6; Rad18 homolog; RHC18; SMC protein 6; Smc6; SMC-6; SMC6 structural maintenance of chromosomes 6-like 1; Smc6l1; structural maintenance of chromosomes 6; Structural maintenance of chromosomes protein 6; YLR383W
Gene Symbols SMC6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SMC6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79677
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.