Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMCC Monoclonal Antibody (6-F6)
GREENER_CHOICE

Catalog No. PIMA544606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544606 Supplier Invitrogen™ Supplier No. MA544606
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Application notes: Indirect ELISA.

Succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate (SMCC) is a non-cleavable and membrane permeable crosslinker. It contains an amine-reactive N -hydroxysuccinimide (NHS ester) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of SMCC and Sulfo-SMCC and are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCC
Applications ELISA
Classification Monoclonal
Clone 6-F6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias Succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate
Host Species Mouse
Immunogen Recombinant protein fused with SMCC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.